siRNA Details
Gene Symbol: TMEM121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025268.2 | NP_079544.1 | 2 | 193 | ||
XM_005268101.3 | XP_005268158.1 | 2 | 800 | |||
XM_006720261.3 | XP_006720324.1 | 2 | 632 | |||
GenBank mRNA | BC004221.2 | - | 2 | 193 | 3530864 |
