siRNA Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030576.3 | NP_085053.1 | 5 | 502 | ||
| XM_005257703.1 | XP_005257760.1 | 5 | 433 | |||
| XM_005257705.3 | XP_005257762.1 | 5 | 613 | |||
| XM_006722124.1 | XP_006722187.1 | 5 | 409 | |||
| GenBank mRNA | AK092301.1 | - | 5 | 468 | ||
| AK307353.1 | - | 3 | 478 | |||
| AK310718.1 | - | 2 | 564 | |||
| AY037154.1 | - | 5 | 538 | |||
| BC004400.1 | - | 5 | 483 | 3636469 | ||
| BC051812.1 | - | 5 | 738 | 5732102 | ||

LIMD2
Genomic Map