siRNA Details
| Gene Symbol: CYB5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030579.2 | NP_085056.2 | 1 | 187 | ||
| GenBank mRNA | AB009282.1 | - | 1 | 214 | ||
| AK291576.1 | - | 1 | 119 | |||
| BC004373.1 | - | 1 | 122 | 3639133 | ||
| BC014431.1 | - | 1 | 145 | 3542294 | ||
| BX647922.1 | - | 1 | 163 | |||

CYB5B
Genomic Map