siRNA Details
| Gene Symbol: ASXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030632.2 | NP_085135.1 | 11 | 1334 | ||
| XM_005258356.1 | XP_005258413.1 | 12 | 1307 | |||
| XM_011526205.2 | XP_011524507.1 | 12 | 8097 | |||
| XM_011526206.2 | XP_011524508.1 | 12 | 4862 | |||
| XM_011526209.1 | XP_011524511.1 | 13 | 1491 | |||
| XM_011526210.1 | XP_011524512.1 | 13 | 1548 | |||
| XM_011526212.1 | XP_011524514.1 | 12 | 1452 | |||
| XM_011526213.1 | XP_011524515.1 | 12 | 1545 | |||
| XM_017026012.1 | XP_016881501.1 | 12 | 4709 | |||
| XM_017026013.1 | XP_016881502.1 | 13 | 5071 | |||
| XM_017026014.1 | XP_016881503.1 | 13 | 1508 | |||
| GenBank mRNA | AK131454.1 | - | 13 | 1377 | ||

ASXL3
Genomic Map