siRNA Details
| Gene Symbol: GRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080423.3 | NP_001073892.3 | 10, 11 | 1197 | ||
| XM_011534141.2 | XP_011532443.1 | 10, 11 | 1279 | |||
| XM_011534142.2 | XP_011532444.1 | 10, 11 | 1191 | |||
| XM_011534145.2 | XP_011532447.1 | 4, 5 | 925 | |||
| XM_011534146.2 | XP_011532448.1 | 9, 10 | 1758 | |||
| GenBank mRNA | AB051506.2 | - | 10, 11 | 1183 | ||

GRIP2
Genomic Map