siRNA Details
| Gene Symbol: KDM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030647.1 | NP_085150.1 | 15, 16 | 2009 | ||
| XM_011516587.2 | XP_011514889.1 | 9, 10 | 1034 | |||
| GenBank mRNA | AB051505.1 | - | 3, 4 | 314 | ||
| AK131497.1 | - | 15, 16 | 2010 | |||
| BX641017.1 | - | 7, 8 | 789 | |||
| BX649115.1 | - | 10, 11 | 1276 | |||

KDM7A
Genomic Map