siRNA Details
| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF032109.2 | - | 3 | 2851 | ||
| AF032110.1 | - | 3 | 689 | |||
| BX648746.1 | - | 3 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRD1ASP| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF032109.2 | - | 3 | 2851 | ||
| AF032110.1 | - | 3 | 689 | |||
| BX648746.1 | - | 3 | 340 | |||