siRNA Details
| Gene Symbol: TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145549.2 | NP_001139021.1 | 10 | 1275 | ||
| NM_030810.3 | NP_110437.2 | 10 | 1311 | |||
| GenBank mRNA | AJ440721.1 | - | 10 | 1066 | ||
| AK025006.1 | - | 4 | 1051 | |||
| AK075291.1 | - | 10 | 1285 | |||
| AK098089.1 | - | 13 | 1287 | |||
| AK315598.1 | - | 10 | 1276 | |||
| AL833008.1 | - | 10 | 1155 | |||
| AL834423.1 | - | 7 | 709 | |||
| AY326464.1 | - | 10 | 1299 | |||
| AY358646.1 | - | 10 | 1305 | |||
| BC001199.1 | - | 10 | 1030 | 3356029 | ||
| Gene Symbol: BLOC1S5-TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037616.1 | 13 | 1432 | |||
| GenBank mRNA | BC052310.1 | - | 13 | 1382 | 6671945 | |

TXNDC5
Genomic Map