siRNA Details
| Gene Symbol: NETO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201465.1 | NP_001188394.1 | 3 | 484 | ||
| NM_138966.3 | NP_620416.1 | 3 | 484 | |||
| NM_138999.1 | NP_620552.1 | 3 | 299 | |||
| XM_005266774.4 | XP_005266831.1 | 3 | 422 | |||
| XM_017026017.1 | XP_016881506.1 | 3 | 346 | |||
| XM_017026018.1 | XP_016881507.1 | 3 | 346 | |||
| XM_017026019.1 | XP_016881508.1 | 3 | 506 | |||
| XM_017026020.1 | XP_016881509.1 | 3 | 510 | |||
| XM_017026021.1 | XP_016881510.1 | 2 | 172 | |||
| XM_017026022.1 | XP_016881511.1 | 3 | 264 | |||
| XM_017026023.1 | XP_016881512.1 | 3 | 264 | |||
| XM_017026024.1 | XP_016881513.1 | 3 | 479 | |||
| GenBank mRNA | AF448838.1 | - | 3 | 490 | ||
| AF448839.1 | - | 3 | 299 | |||
| AL834354.1 | - | 1 | 103 | |||
| BC050329.1 | - | 3 | 486 | 4821317 | ||

NETO1
Genomic Map