siRNA Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030981.2 | NP_112243.1 | 6 | 1777 | ||
| XM_017018378.1 | XP_016873867.1 | 5 | 1944 | |||
| GenBank mRNA | AF092437.1 | - | 7 | 861 | ||
| AJ245875.1 | - | 5 | 1682 | |||
| AL136635.1 | - | 6 | 1729 | |||
| BC028331.1 | - | 6 | 1726 | |||
| BC071169.1 | - | 6 | 1716 | 6026121 | ||

RAB1B
Genomic Map