siRNA Details
| Gene Symbol: CD99L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184808.1 | NP_001171737.1 | 6 | 556 | ||
| NM_001242614.1 | NP_001229543.1 | 9 | 787 | |||
| NM_031462.3 | NP_113650.2 | 9 | 775 | |||
| NM_134445.3 | NP_604394.1 | 6 | 559 | |||
| NM_134446.3 | NP_604395.1 | 7 | 628 | |||
| XM_017029890.1 | XP_016885379.1 | 9 | 639 | |||
| XM_017029891.1 | XP_016885380.1 | 7 | 626 | |||
| GenBank mRNA | AK023776.1 | - | 8 | 461 | ||
| AK024278.1 | - | 5 | 282 | |||
| AK125020.1 | - | 9 | 598 | |||
| AK290200.1 | - | 9 | 551 | |||
| AK291375.1 | - | 7 | 441 | |||
| AK293244.1 | - | 6 | 362 | |||
| AL136580.1 | - | 9 | 664 | |||
| AY078165.1 | - | 9 | 583 | |||
| AY078166.1 | - | 7 | 436 | |||
| AY078167.1 | - | 6 | 367 | |||
| AY358837.1 | - | 9 | 648 | |||
| BC025729.1 | - | 10 | 1996 | 5203087 | ||
| BC030536.1 | - | 9 | 635 | 5242536 | ||
| KU178797.1 | - | 9 | 547 | |||
| KU178798.1 | - | 3 | 118 | |||

CD99L2
Genomic Map