siRNA Details
| Gene Symbol: PRSS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318395.1 | NP_001305324.1 | 5 | 1038 | ||
| NM_031948.4 | NP_114154.1 | 6 | 1201 | |||
| XM_011522689.2 | XP_011520991.1 | 4 | 930 | |||
| GenBank mRNA | AB056161.1 | - | 6 | 860 | ||
| AJ306593.1 | - | 6 | 829 | |||
| AK055576.1 | - | 3 | 1802 | |||
| AY030095.1 | - | 6 | 810 | |||
| AY359106.1 | - | 6 | 845 | |||
| BC034294.2 | - | 6 | 865 | 4752547 | ||
| BC036912.2 | - | 5 | 1049 | |||

PRSS27
Genomic Map