siRNA Details
| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032028.3 | NP_114417.1 | 1 | 690 | ||
| GenBank mRNA | AK313332.1 | - | 1 | 643 | ||
| AY028964.1 | - | 1 | 588 | |||
| BC022515.1 | - | 1 | 628 | 4816988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSK1B| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032028.3 | NP_114417.1 | 1 | 690 | ||
| GenBank mRNA | AK313332.1 | - | 1 | 643 | ||
| AY028964.1 | - | 1 | 588 | |||
| BC022515.1 | - | 1 | 628 | 4816988 | ||