siRNA Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032164.2 | NP_115540.2 | 1 | 598 | ||
| XM_017012710.1 | XP_016868199.1 | 1 | 646 | |||
| XM_017012711.1 | XP_016868200.1 | 1 | 646 | |||
| GenBank mRNA | AK022360.1 | - | 1 | 641 | ||
| AK172865.1 | - | 2 | 519 | |||
| AK222948.1 | - | 1 | 591 | |||
| AY261374.1 | - | 1 | 641 | |||
| AY642122.1 | - | 1 | 625 | |||
| BC017051.1 | - | 1 | 608 | |||
| BC025241.1 | - | 1 | 625 | 4641268 | ||
| BC051847.1 | - | 1 | 598 | 6007172 | ||

ZNF394
Genomic Map