siRNA Details
| Gene Symbol: CEP78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098802.1 | NP_001092272.1 | 2, 3 | 698 | ||
| NM_032171.1 | NP_115547.1 | 2, 3 | 698 | |||
| XM_005252263.4 | XP_005252320.1 | 2, 3 | 711 | |||
| XM_005252265.2 | XP_005252322.1 | 2, 3 | 711 | |||
| XM_005252266.4 | XP_005252323.1 | 2, 3 | 705 | |||
| XM_017015192.1 | XP_016870681.1 | 2, 3 | 711 | |||
| XM_017015193.1 | XP_016870682.1 | 2, 3 | 711 | |||
| XM_017015194.1 | XP_016870683.1 | 2, 3 | 711 | |||
| XM_017015195.1 | XP_016870684.1 | 2, 3 | 711 | |||
| XM_017015196.1 | XP_016870685.1 | 2, 3 | 705 | |||
| GenBank mRNA | AK022705.1 | - | 2, 3 | 678 | ||
| AK303457.1 | - | 2, 3 | 748 | |||
| BC058931.1 | - | 2, 3 | 611 | |||
| BC128058.1 | - | 2, 3 | 566 | 40078333 | ||

CEP78
Genomic Map