siRNA Details
Gene Symbol: KIAA1109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015312.3 | NP_056127.2 | 76 | 13420 | ||
XM_005263282.1 | XP_005263339.1 | 79 | 13811 | |||
XM_005263287.1 | XP_005263344.1 | 78 | 13748 | |||
XM_006714344.1 | XP_006714407.1 | 78 | 13745 | |||
XM_011532319.1 | XP_011530621.1 | 80 | 14012 | |||
XM_011532320.2 | XP_011530622.1 | 81 | 14107 | |||
XM_011532321.2 | XP_011530623.1 | 79 | 13814 | |||
XM_011532322.1 | XP_011530624.1 | 80 | 14009 | |||
XM_011532323.1 | XP_011530625.1 | 79 | 13961 | |||
XM_011532324.1 | XP_011530626.1 | 79 | 13949 | |||
XM_011532325.1 | XP_011530627.1 | 80 | 13907 | |||
XM_011532326.1 | XP_011530628.1 | 79 | 13883 | |||
XM_011532327.1 | XP_011530629.1 | 81 | 14058 | |||
XM_011532330.1 | XP_011530632.1 | 64 | 11958 | |||
XM_017008695.1 | XP_016864184.1 | 79 | 13706 | |||
XM_017008696.1 | XP_016864185.1 | 78 | 13655 | |||
XM_017008697.1 | XP_016864186.1 | 78 | 13643 | |||
XM_017008698.1 | XP_016864187.1 | 58 | 11974 | |||
GenBank mRNA | AB029032.2 | - | 42 | 7619 | ||
AK127698.1 | - | 7 | 1200 | |||
AK303483.1 | - | 4 | 648 | |||
AL137532.1 | - | 4 | 643 | |||
AL833461.1 | - | 16 | 2500 | |||
DQ335469.1 | - | 76 | 13420 |
