siRNA Details
Gene Symbol: KIAA1109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015312.3 | NP_056127.2 | 27 | 4584 | ||
XM_005263282.1 | XP_005263339.1 | 29 | 4912 | |||
XM_005263287.1 | XP_005263344.1 | 29 | 4912 | |||
XM_006714344.1 | XP_006714407.1 | 29 | 4909 | |||
XM_011532319.1 | XP_011530621.1 | 29 | 4912 | |||
XM_011532320.2 | XP_011530622.1 | 30 | 5007 | |||
XM_011532321.2 | XP_011530623.1 | 28 | 4714 | |||
XM_011532322.1 | XP_011530624.1 | 29 | 4909 | |||
XM_011532323.1 | XP_011530625.1 | 29 | 4912 | |||
XM_011532324.1 | XP_011530626.1 | 29 | 4912 | |||
XM_011532325.1 | XP_011530627.1 | 29 | 4912 | |||
XM_011532326.1 | XP_011530628.1 | 28 | 4783 | |||
XM_011532327.1 | XP_011530629.1 | 30 | 4958 | |||
XM_011532330.1 | XP_011530632.1 | 13 | 2858 | |||
XM_017008695.1 | XP_016864184.1 | 29 | 4912 | |||
XM_017008696.1 | XP_016864185.1 | 29 | 4912 | |||
XM_017008697.1 | XP_016864186.1 | 29 | 4912 | |||
XM_017008698.1 | XP_016864187.1 | 7 | 2874 | |||
XM_017008699.1 | XP_016864188.1 | 29 | 4912 | |||
GenBank mRNA | CR936613.1 | - | 1 | 257 | ||
DQ335469.1 | - | 27 | 4584 |
