siRNA Details
Gene Symbol: KIAA1109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015312.3 | NP_056127.2 | 35 | 5836 | ||
XM_005263282.1 | XP_005263339.1 | 37 | 6164 | |||
XM_005263287.1 | XP_005263344.1 | 37 | 6164 | |||
XM_006714344.1 | XP_006714407.1 | 37 | 6161 | |||
XM_011532319.1 | XP_011530621.1 | 37 | 6164 | |||
XM_011532320.2 | XP_011530622.1 | 38 | 6259 | |||
XM_011532321.2 | XP_011530623.1 | 36 | 5966 | |||
XM_011532322.1 | XP_011530624.1 | 37 | 6161 | |||
XM_011532323.1 | XP_011530625.1 | 37 | 6164 | |||
XM_011532324.1 | XP_011530626.1 | 37 | 6164 | |||
XM_011532325.1 | XP_011530627.1 | 37 | 6164 | |||
XM_011532326.1 | XP_011530628.1 | 36 | 6035 | |||
XM_011532327.1 | XP_011530629.1 | 38 | 6210 | |||
XM_011532330.1 | XP_011530632.1 | 21 | 4110 | |||
XM_017008695.1 | XP_016864184.1 | 37 | 6164 | |||
XM_017008696.1 | XP_016864185.1 | 37 | 6164 | |||
XM_017008697.1 | XP_016864186.1 | 37 | 6164 | |||
XM_017008698.1 | XP_016864187.1 | 15 | 4126 | |||
XM_017008699.1 | XP_016864188.1 | 37 | 6164 | |||
GenBank mRNA | AB029032.2 | - | 1 | 35 | ||
AK095216.1 | - | 4 | 545 | |||
AK309063.1 | - | 7 | 957 | |||
CR936613.1 | - | 9 | 1509 | |||
DQ335469.1 | - | 35 | 5836 |
