siRNA Details
| Gene Symbol: FBXW9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032301.2 | NP_115677.2 | 9 | 1345 | ||
| XM_005260096.4 | XP_005260153.1 | 8 | 1464 | |||
| GenBank mRNA | AK123037.1 | - | 9 | 1344 | ||
| AK302744.1 | - | 7 | 1070 | |||
| AW083348.1 | - | 2 | 405 | |||
| AY736034.1 | - | 9 | 1345 | |||
| BC004290.1 | - | 9 | 1322 | |||

FBXW9
Genomic Map