siRNA Details
| Gene Symbol: COQ5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032314.3 | NP_115690.3 | 3 | 438 | ||
| XM_006719639.2 | XP_006719702.1 | 4 | 538 | |||
| GenBank mRNA | AK222610.1 | - | 3 | 428 | ||
| AK225242.1 | - | 3 | 428 | |||
| AK293656.1 | - | 3 | 459 | |||
| AK315681.1 | - | 3 | 429 | |||
| BC004916.2 | - | 3 | 414 | |||
| BC107874.1 | - | 3 | 432 | 6452052 | ||
| BP307378.1 | - | 3 | 438 | |||
| BX647562.1 | - | 3 | 427 | |||
| EU700459.1 | - | 3 | 432 | |||

COQ5
Genomic Map