siRNA Details
| Gene Symbol: NUDT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128612.2 | NP_001122084.1 | 6 | 856 | ||
| NM_001128613.2 | NP_001122085.1 | 5 | 871 | |||
| NM_001271831.1 | NP_001258760.1 | 5 | 757 | |||
| NM_032344.3 | NP_115720.1 | 6 | 970 | |||
| GenBank mRNA | BC006129.1 | - | 6 | 954 | 3625545 | |
| BM923060.1 | - | 5 | 756 | |||

NUDT22
Genomic Map