siRNA Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138418.2 | NP_612427.2 | 4, 5 | 554 | ||
| GenBank mRNA | AF386504.1 | - | 3, 4 | 426 | ||
| AK054811.1 | - | 2, 3 | 1950 | |||
| BC001912.1 | - | 4, 5 | 554 | 3546959 | ||
| BC007346.2 | - | 3, 4 | 346 | |||
| BC111391.1 | - | 4, 5 | 456 | 6457301 | ||

MCRIP2
Genomic Map