siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052907.3 | NP_443139.2 | 2 | 86 | ||
| XM_011537852.2 | XP_011536154.1 | 2 | 143 | |||
| XM_017018767.1 | XP_016874256.1 | 3 | 252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM132B| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052907.3 | NP_443139.2 | 2 | 86 | ||
| XM_011537852.2 | XP_011536154.1 | 2 | 143 | |||
| XM_017018767.1 | XP_016874256.1 | 3 | 252 | |||