siRNA Details
| Gene Symbol: NKX6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177400.2 | NP_796374.1 | 1 | 149 | ||
| XM_017016789.1 | XP_016872278.1 | 1 | 143 | |||
| GenBank mRNA | BC101635.1 | - | 1 | 149 | 8069141 | |
| BC104823.1 | - | 1 | 149 | 8143826 | ||

NKX6-2
Genomic Map