siRNA Details
| Gene Symbol: GON7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032490.4 | NP_115879.2 | 2 | 415 | ||
| GenBank mRNA | AF277185.1 | - | 2 | 160 | ||
| BC021733.2 | - | 2 | 395 | 4796574 | ||
| BC065216.1 | - | 2 | 317 | |||
| BC107429.1 | - | 2 | 383 | 4809642 | ||
| BC109390.1 | - | 2 | 326 | |||

GON7
Genomic Map