siRNA Details
| Gene Symbol: GOLGA2P2Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001555.2 | 7, 8 | 879 | |||
| GenBank mRNA | AF332229.1 | - | 7, 8 | 878 | ||
| Gene Symbol: GOLGA2P3Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002195.2 | 7, 8 | 879 | |||

GOLGA2P2Y
Genomic Map