siRNA Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032532.2 | NP_115921.2 | 5 | 845 | ||
| XM_011536190.2 | XP_011534492.1 | 4 | 768 | |||
| XM_011536191.2 | XP_011534493.1 | 2 | 486 | |||
| GenBank mRNA | AB058769.2 | - | 4 | 520 | ||
| BC146783.1 | - | 4 | 520 | 9007239 | ||
| BC150607.1 | - | 4 | 520 | 9056977 | ||
| DQ009660.1 | - | 5 | 845 | |||

FNDC1
Genomic Map