siRNA Details
| Gene Symbol: DSCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026838.1 | 4 | 683 | |||
| NR_026839.1 | 3 | 586 | ||||
| NR_026840.1 | 4 | 444 | ||||
| NR_026841.1 | 4 | 426 | ||||
| NR_026842.1 | 3 | 329 | ||||
| GenBank mRNA | AA707621.1 | - | 1 | 131 | ||
| AA770014.1 | - | 1 | 137 | |||
| AF321193.1 | - | 4 | 401 | |||
| AF426256.1 | - | 4 | 349 | |||
| AJ001904.1 | - | 4 | 386 | |||
| AJ297914.2 | - | 4 | 404 | |||
| AJ301615.2 | - | 3 | 307 | |||
| AJ305085.2 | - | 3 | 564 | |||
| AJ306839.2 | - | 4 | 422 | |||
| AJ306840.2 | - | 4 | 476 | |||
| AJ783421.1 | - | 4 | 661 | |||
| BC015981.1 | - | 3 | 311 | |||
| BC029353.1 | - | 3 | 586 | |||
| BC070229.1 | - | 3 | 288 | |||
| BC117304.1 | - | 4 | 358 | |||
| BC117330.1 | - | 4 | 358 | |||
| BC143995.1 | - | 4 | 376 | |||
| BG189423.1 | - | 1 | 81 | |||

DSCR8
Genomic Map