siRNA Details
| Gene Symbol: KDM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005366.1 | NP_001005366.1 | 13 | 2188 | ||
| NM_032590.4 | NP_115979.3 | 13 | 1978 | |||
| XM_005253955.3 | XP_005254012.1 | 13 | 2188 | |||
| XM_005253956.3 | XP_005254013.1 | 13 | 1785 | |||
| XM_005253961.4 | XP_005254018.1 | 2 | 273 | |||
| XM_011538867.2 | XP_011537169.1 | 13 | 3231 | |||
| XM_011538868.2 | XP_011537170.1 | 13 | 3227 | |||
| XM_011538869.2 | XP_011537171.1 | 13 | 3229 | |||
| XM_011538875.2 | XP_011537177.1 | 2 | 273 | |||
| GenBank mRNA | AB031230.1 | - | 3 | 437 | ||
| AJ459424.1 | - | 13 | 1950 | |||
| AK027440.1 | - | 3 | 367 | |||
| AK127328.1 | - | 13 | 2188 | |||
| AK299827.1 | - | 2 | 283 | |||
| AK316470.1 | - | 2 | 285 | |||
| AL133572.1 | - | 9 | 1491 | |||
| BC115379.1 | - | 12 | 1838 | |||
| BC115380.1 | - | 13 | 1917 | 40029132 | ||

KDM2B
Genomic Map