siRNA Details
| Gene Symbol: PERM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291366.1 | NP_001278295.1 | 4 | 2374 | ||
| NM_001291367.1 | NP_001278296.1 | 5 | 2021 | |||
| XM_017002583.1 | XP_016858072.1 | 4 | 2476 | |||
| XM_017002584.1 | XP_016858073.1 | 4 | 2499 | |||
| XM_017002585.1 | XP_016858074.1 | 3 | 2865 | |||
| GenBank mRNA | AK123855.1 | - | 5 | 1997 | ||
| AK312221.1 | - | 2 | 130 | |||
| BC006300.2 | - | 2 | 171 | |||
| KF150175.1 | - | 4 | 2374 | |||

PERM1
Genomic Map