siRNA Details
| Gene Symbol: PLPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032728.3 | NP_116117.3 | 1 | 537 | ||
| XM_005272230.3 | XP_005272287.1 | 1 | 549 | |||
| XM_005272231.1 | XP_005272288.1 | 1 | 549 | |||
| GenBank mRNA | AK027568.1 | - | 1 | 491 | ||
| AK075207.1 | - | 1 | 537 | |||
| AK315502.1 | - | 1 | 366 | |||
| BC006362.2 | - | 1 | 494 | 4129897 | ||

PLPP7
Genomic Map