siRNA Details
| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026827.1 | 2 | 459 | |||
| GenBank mRNA | AK090789.1 | - | 2 | 406 | ||
| AK092104.1 | - | 2 | 409 | |||
| AK293138.1 | - | 2 | 459 | |||
| BC007394.2 | - | 2 | 412 | |||
| BI517331.1 | - | 2 | 203 | |||
| BM738658.1 | - | 2 | 442 | |||

LINC00839
Genomic Map