siRNA Details
| Gene Symbol: MPND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159846.2 | NP_001153318.1 | 3 | 482 | ||
| NM_001300862.1 | NP_001287791.1 | 3 | 482 | |||
| NM_032868.5 | NP_116257.2 | 3 | 482 | |||
| XM_006722926.2 | XP_006722989.1 | 3 | 443 | |||
| GenBank mRNA | AK027887.1 | - | 3 | 455 | ||
| AK308468.1 | - | 3 | 443 | |||
| BC032652.1 | - | 3 | 439 | 5503607 | ||
| BG696748.1 | - | 1 | 68 | |||
| BP266054.1 | - | 3 | 442 | |||
| BU150675.1 | - | 3 | 429 | |||
| BX462788.2 | - | 3 | 482 | |||

MPND
Genomic Map