siRNA Details
| Gene Symbol: MPND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300862.1 | NP_001287791.1 | 7, 8 | 985 | ||
| NM_032868.5 | NP_116257.2 | 7, 8 | 985 | |||
| XM_006722926.2 | XP_006722989.1 | 7, 8 | 946 | |||
| GenBank mRNA | AK308468.1 | - | 7, 8 | 946 | ||
| BC032652.1 | - | 7, 8 | 942 | 5503607 | ||
| CR977937.1 | - | 4, 5 | 322 | |||

MPND
Genomic Map