siRNA Details
| Gene Symbol: FAM83A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288587.2 | NP_001275516.1 | 3 | 1180 | ||
| NM_032899.6 | NP_116288.2 | 5 | 1772 | |||
| NM_207006.2 | NP_996889.1 | 5 | 2857 | |||
| XM_005251087.2 | XP_005251144.1 | 3 | 977 | |||
| GenBank mRNA | AF497803.1 | - | 4 | 1097 | ||
| AK095301.1 | - | 5 | 2857 | |||
| AK098407.1 | - | 4 | 446 | |||
| BC007828.2 | - | 3 | 851 | 3610547 | ||
| BC052300.1 | - | 4 | 1161 | 6421272 | ||
| DQ280322.1 | - | 3 | 890 | |||
| DQ280323.1 | - | 5 | 1745 | |||

FAM83A
Genomic Map