siRNA Details
| Gene Symbol: PYURF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032906.4 | NP_116295.1 | 2 | 334 | ||
| GenBank mRNA | AK312082.1 | - | 2 | 309 | ||
| BG706896.1 | - | 2 | 339 | |||
| CV812938.1 | - | 2 | 270 | |||
| Gene Symbol: PIGY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042616.2 | NP_001036081.1 | 2 | 334 | ||
| GenBank mRNA | AB206972.1 | - | 2 | 306 | ||
| BC007876.2 | - | 2 | 306 | 3947944 | ||
| BI548853.1 | - | 2 | 339 | |||

PYURF
Genomic Map