siRNA Details
| Gene Symbol: TMEM60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032936.3 | NP_116325.1 | 2 | 328 | ||
| GenBank mRNA | AF260336.1 | - | 2 | 329 | ||
| BC065930.1 | - | 2 | 252 | 6146561 | ||
| BX117806.1 | - | 2 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM60| Gene Symbol: TMEM60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032936.3 | NP_116325.1 | 2 | 328 | ||
| GenBank mRNA | AF260336.1 | - | 2 | 329 | ||
| BC065930.1 | - | 2 | 252 | 6146561 | ||
| BX117806.1 | - | 2 | 328 | |||