siRNA Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024127.1 | 1 | 230 | |||
| GenBank mRNA | AF277181.1 | - | 18 | |||
| AK092096.1 | - | 1 | 251 | |||
| BC034611.1 | - | 1 | 40 | |||
| BC045792.1 | - | 1 | 248 | |||
| BC064589.1 | - | 1 | 218 | |||
| BC068048.1 | - | 1 | 230 | |||
| KJ782215.1 | - | 2 | 1359 | |||

SNHG12
Genomic Map