siRNA Details
| Gene Symbol: KRTAP4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033060.2 | NP_149049.1 | 1 | 85 | ||
| GenBank mRNA | AJ406942.1 | - | 1 | 85 | ||
| BC065372.1 | - | 2 | 582 | |||
| BC103845.1 | - | 1 | 65 | 40002744 | ||
| BC103846.2 | - | 1 | 65 | 40002745 | ||

KRTAP4-1
Genomic Map