siRNA Details
| Gene Symbol: KIAA1644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099294.1 | NP_001092764.1 | 4 | 433 | ||
| XM_005261790.3 | XP_005261847.1 | 4 | 531 | |||
| GenBank mRNA | AB051431.1 | - | 2 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1644| Gene Symbol: KIAA1644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099294.1 | NP_001092764.1 | 4 | 433 | ||
| XM_005261790.3 | XP_005261847.1 | 4 | 531 | |||
| GenBank mRNA | AB051431.1 | - | 2 | 99 | ||