siRNA Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033017.3 | NP_148977.2 | 3 | 715 | ||
| NM_033091.2 | NP_149082.1 | 2 | 637 | |||
| GenBank mRNA | AF220023.1 | - | 4 | 652 | ||
| AF220024.1 | - | 2 | 574 | |||
| AK092892.1 | - | 2 | 599 | |||
| AK293565.1 | - | 4 | 491 | |||
| BC011763.2 | - | 2 | 530 | 3617674 | ||
| BC025949.1 | - | 2 | 530 | 4620469 | ||

TRIM4
Genomic Map