siRNA Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033017.3 | NP_148977.2 | 4 | 836 | ||
| NM_033091.2 | NP_149082.1 | 3 | 758 | |||
| GenBank mRNA | AF220023.1 | - | 5 | 773 | ||
| AF220024.1 | - | 3 | 695 | |||
| AK092892.1 | - | 3 | 720 | |||
| AK293565.1 | - | 5 | 612 | |||
| BC011763.2 | - | 3 | 651 | 3617674 | ||
| BC025949.1 | - | 3 | 651 | 4620469 | ||

TRIM4
Genomic Map