siRNA Details
| Gene Symbol: ZNF628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033113.2 | NP_149104.3 | 3 | 2950 | ||
| XM_005259371.3 | XP_005259428.1 | 2 | 2595 | |||
| GenBank mRNA | AF367249.1 | - | 1 | 12 | ||
| BC047332.1 | - | 1 | 108 | |||
| BC089449.1 | - | 3 | 2950 | 30560806 | ||

ZNF628
Genomic Map