siRNA Details
| Gene Symbol: FER1L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293083.1 | NP_001280012.1 | 42 | 4663 | ||
| XM_011512110.2 | XP_011510412.1 | 42 | 4680 | |||
| XM_011512111.2 | XP_011510413.1 | 42 | 4677 | |||
| XM_011512113.2 | XP_011510415.1 | 42 | 4674 | |||
| XM_011512114.2 | XP_011510416.1 | 42 | 4677 | |||
| XM_011512115.2 | XP_011510417.1 | 41 | 4611 | |||
| XM_011512116.2 | XP_011510418.1 | 42 | 4593 | |||
| XM_011512117.2 | XP_011510419.1 | 41 | 4575 | |||
| XM_011512118.2 | XP_011510420.1 | 41 | 4560 | |||
| XM_011512119.2 | XP_011510421.1 | 41 | 4518 | |||
| XM_011512120.2 | XP_011510422.1 | 41 | 4571 | |||
| XM_011512122.2 | XP_011510424.1 | 42 | 4681 | |||
| XM_011512124.2 | XP_011510426.1 | 26 | 4264 | |||
| XM_017005222.1 | XP_016860711.1 | 42 | 4677 | |||
| XM_017005223.1 | XP_016860712.1 | 25 | 3328 | |||
| XM_017005224.1 | XP_016860713.1 | 16 | 2120 | |||
| GenBank mRNA | AL133022.1 | - | 24 | 4066 | ||
| AY461813.1 | - | 41 | 4692 | |||
| BC117324.1 | - | 8 | 871 | |||
| BC126368.1 | - | 8 | 874 | |||
| BC143825.1 | - | 8 | 871 | |||

FER1L5
Genomic Map