siRNA Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182491.2 | NP_872297.2 | 4 | 516 | ||
| XM_017012774.1 | XP_016868263.1 | 4 | 516 | |||
| GenBank mRNA | AI435373.1 | - | 2 | 388 | ||
| AK027118.1 | - | 4 | 458 | |||
| AK290170.1 | - | 4 | 475 | |||
| BC016963.1 | - | 4 | 451 | |||
| BC029558.1 | - | 4 | 459 | 5263788 | ||

ZFAND2A
Genomic Map