siRNA Details
| Gene Symbol: FAM104B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166699.1 | NP_001160171.1 | 3 | 217 | ||
| NM_001166700.1 | NP_001160172.1 | 3 | 217 | |||
| NM_001166701.1 | NP_001160173.1 | 3 | 214 | |||
| NM_001166702.1 | NP_001160174.1 | 3 | 315 | |||
| NM_001166703.1 | NP_001160175.1 | 3 | 393 | |||
| NM_001166704.1 | NP_001160176.1 | 3 | 221 | |||
| NM_138362.3 | NP_612371.2 | 3 | 214 | |||
| NR_030722.1 | 3 | 221 | ||||
| XM_017029951.1 | XP_016885440.1 | 3 | 238 | |||
| XM_017029952.1 | XP_016885441.1 | 3 | 451 | |||
| XM_017029953.1 | XP_016885442.1 | 3 | 311 | |||
| GenBank mRNA | AK000441.1 | - | 3 | 198 | ||
| AK299935.1 | - | 3 | 302 | |||
| AK311333.1 | - | 2 | 102 | |||
| AK311699.1 | - | 3 | 208 | |||
| BC000919.2 | - | 3 | 192 | 3449838 | ||
| BC006406.2 | - | 1 | 176 | |||
| BC017571.1 | - | 3 | 202 | 4105917 | ||
| BX100076.1 | - | 3 | 393 | |||
| CK004182.1 | - | 3 | 211 | |||
| DB216586.1 | - | 3 | 214 | |||
| DC413808.1 | - | 3 | 315 | |||

FAM104B
Genomic Map