siRNA Details
Gene Symbol: PAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013435.2 | NP_001013453.1 | 5 | 509 | ||
NM_130467.4 | NP_569734.2 | 5 | 708 | |||
GenBank mRNA | AJ344352.1 | - | 5 | 678 | ||
BC009230.2 | - | 5 | 697 | 3955765 | ||
BC110803.1 | - | 5 | 443 | 3922538 | ||
BQ437151.1 | - | 5 | 497 |
Gene Symbol: LOC100421746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130179.1 | 5 | 471 | |||
GenBank mRNA | AI651025.1 | - | 1 | 36 |
