siRNA Details
| Gene Symbol: IGHV5-78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF085654.1 | - | 155 | |||
| BC002792.2 | - | 2 | 208 | 3621362 | ||
| X58398.1 | - | 2 | 206 | |||
| X58399.1 | - | 2 | 189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGHV5-78| Gene Symbol: IGHV5-78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF085654.1 | - | 155 | |||
| BC002792.2 | - | 2 | 208 | 3621362 | ||
| X58398.1 | - | 2 | 206 | |||
| X58399.1 | - | 2 | 189 | |||