siRNA Details
Gene Symbol: IGHV5-78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002792.2 | - | 2 | 650 | 3621362 | |
X58398.1 | - | 2 | 648 | |||
X58399.1 | - | 2 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IGHV5-78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002792.2 | - | 2 | 650 | 3621362 | |
X58398.1 | - | 2 | 648 | |||
X58399.1 | - | 2 | 631 |