siRNA Details
| Gene Symbol: ZNF251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138367.1 | NP_612376.1 | 5 | 1902 | ||
| GenBank mRNA | AK000435.1 | - | 5 | 1786 | ||
| AK027286.1 | - | 1 | 938 | |||
| BC006258.2 | - | 1 | 513 | |||
| BC093984.1 | - | 1 | 422 | |||
| BC112137.1 | - | 1 | 422 | |||

ZNF251
Genomic Map